Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IRF6
Entrez Gene ID
614253
Gene Name
interferon regulatory factor 6
Gene Aliases
-
Chromosome Location
Chr.16: 73502286 - 73518532 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076934.1 NP_001070402.1 1 - 2 44 66
GenBank mRNA BC123806.1 - 1 - 2 44 66

View Full Product Details