Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXL8
Entrez Gene ID
614765
Gene Name
F-box and leucine rich repeat protein 8
Gene Aliases
-
Chromosome Location
Chr.18: 34798639 - 34802590 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076937.1 NP_001070405.1 2 - 3 479 74
GenBank mRNA BC123762.1 - 2 - 3 479 74

View Full Product Details