Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABRR2
Entrez Gene ID
522099
Gene Name
gamma-aminobutyric acid type A receptor subunit rho2
Gene Aliases
-
Chromosome Location
Chr.9: 60845380 - 60885598 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077946.1 NP_001071414.2 4 - 5 648 84
GenBank mRNA BC122770.1 - 4 - 5 648 84

View Full Product Details