Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDSN
Entrez Gene ID
522351
Gene Name
corneodesmosin
Gene Aliases
-
Chromosome Location
Chr.23: 28021400 - 28025617 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077950.1 NP_001071418.1 1 - 2 87 142
GenBank mRNA BC119921.1 - 1 - 2 87 142

View Full Product Details