Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC4H2
Entrez Gene ID
768236
Gene Name
zinc finger C4H2-type containing
Gene Aliases
-
Chromosome Location
Chr.X: 95636741 - 95712628 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078111.1 NP_001071579.1 4 - 5 627 75
GenBank mRNA BC123715.1 - 4 - 5 627 75
BT030574.1 - 4 - 5 582 75

View Full Product Details