Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLPP7
Entrez Gene ID
768245
Gene Name
phospholipid phosphatase 7 (inactive)
Gene Aliases
PPAPDC3
Chromosome Location
Chr.11: 101477749 - 101492970 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078118.1 NP_001071586.1 1 - 2 657 95
GenBank mRNA BC123770.1 - 1 - 2 657 95

View Full Product Details