Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VSIR
Entrez Gene ID
783068
Gene Name
V-set immunoregulatory receptor
Gene Aliases
C28H10orf54
Chromosome Location
Chr.28: 27895738 - 27920748 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080347.1 NP_001073816.1 4 - 5 797 68
GenBank mRNA BC123904.1 - 4 - 5 797 68

View Full Product Details