Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC51B
Entrez Gene ID
507185
Gene Name
solute carrier family 51 subunit beta
Gene Aliases
OSTB, OSTBETA
Chromosome Location
Chr.10: 11979491 - 11993947 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077867.2 NP_001071335.1 2 - 3 260 73
GenBank mRNA BC126778.1 - 2 - 3 261 73

View Full Product Details