Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EOGT
Entrez Gene ID
508782
Gene Name
EGF domain specific O-linked N-acetylglucosamine transferase
Gene Aliases
AER61, C22H3orf64
Chromosome Location
Chr.22: 32533637 - 32575456 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077882.1 NP_001071350.1 3 - 4 634 61
GenBank mRNA BC126625.1 - 3 - 4 634 61

View Full Product Details