Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCNT2
Entrez Gene ID
514131
Gene Name
bucentaur-2
Gene Aliases
P97BCNT2
Chromosome Location
Chr.18: 2756806 - 2799613 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077919.2 NP_001071387.1 9 - 10 2064 79
GenBank mRNA AB259013.1 - 9 - 10 2052 79
BC149202.1 - 9 - 10 2037 79

View Full Product Details