Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C24H18orf21
Entrez Gene ID
512779
Gene Name
chromosome 24 C18orf21 homolog
Gene Aliases
C18orf21
Chromosome Location
Chr.24: 21138186 - 21146243 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079590.2 NP_001073058.1 3 - 4 335 136
GenBank mRNA BC122840.1 - 3 - 4 336 136

View Full Product Details