Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBX15
Entrez Gene ID
521974
Gene Name
T-box transcription factor 15
Gene Aliases
-
Chromosome Location
Chr.3: 24129612 - 24255111 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079775.1 NP_001073243.1 4 - 5 973 74
GenBank mRNA BC126761.1 - 4 - 5 973 74

View Full Product Details