Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C16H1orf21
Entrez Gene ID
781721
Gene Name
chromosome 16 C1orf21 homolog
Gene Aliases
-
Chromosome Location
Chr.16: 65410962 - 65657438 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081547.1 NP_001075016.1 5 - 6 733 105
GenBank mRNA BC133418.1 - 5 - 6 733 105

View Full Product Details