Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD22
Entrez Gene ID
504991
Gene Name
ankyrin repeat domain 22
Gene Aliases
-
Chromosome Location
Chr.26: 10503217 - 10541131 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081579.1 NP_001075048.1 1 - 2 209 86
GenBank mRNA BC133416.1 - 1 - 2 209 86

View Full Product Details