Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAGE4
Entrez Gene ID
505725
Gene Name
PAGE family member 4
Gene Aliases
-
Chromosome Location
Chr.X: 87516566 - 87520757 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081580.2 NP_001075049.1 4 - 5 424 90
GenBank mRNA BC133479.1 - 4 - 5 425 90

View Full Product Details