Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SGO1
Entrez Gene ID
614056
Gene Name
shugoshin 1
Gene Aliases
-
Chromosome Location
Chr.1: 157392690 - 157411178 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081617.1 NP_001075086.1 7 - 8 1535 157
GenBank mRNA BC133396.1 - 7 - 8 1535 157

View Full Product Details