Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CMTR1
Entrez Gene ID
509620
Gene Name
cap methyltransferase 1
Gene Aliases
FTSJD2, MTr1
Chromosome Location
Chr.23: 11318812 - 11372352 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082430.2 NP_001075899.1 14 - 15 1688 75
GenBank mRNA BC133559.1 - 14 - 15 1689 75

View Full Product Details