Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OCLN
Entrez Gene ID
512405
Gene Name
occludin
Gene Aliases
-
Chromosome Location
Chr.20: 10229723 - 10276416 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082433.2 NP_001075902.1 5 - 6 1261 71
GenBank mRNA AY589500.1 - 3 - 4 808 71
BC133617.1 - 5 - 6 1262 71

View Full Product Details