Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNI
Entrez Gene ID
504847
Gene Name
cyclin I
Gene Aliases
-
Chromosome Location
Chr.6: 91881487 - 91920418 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083373.1 NP_001076842.1 3 - 4 832 94
GenBank mRNA BC114754.1 - 3 - 4 832 94

View Full Product Details