Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSD17B7
Entrez Gene ID
505212
Gene Name
hydroxysteroid 17-beta dehydrogenase 7
Gene Aliases
-
Chromosome Location
Chr.3: 6634641 - 6664631 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083375.1 NP_001076844.1 4 - 5 449 86
GenBank mRNA BC114716.1 - 4 - 5 449 86
EF661660.1 - 4 - 5 437 86

View Full Product Details