Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD2BP2
Entrez Gene ID
513857
Gene Name
CD2 cytoplasmic tail binding protein 2
Gene Aliases
-
Chromosome Location
Chr.25: 26539114 - 26544190 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083682.2 NP_001077151.1 6 - 7 972 79
GenBank mRNA BC134720.1 - 6 - 7 973 79

View Full Product Details