Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35F6
Entrez Gene ID
525690
Gene Name
solute carrier family 35 member F6
Gene Aliases
C11H2orf18
Chromosome Location
Chr.11: 72804691 - 72820755 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083704.1 NP_001077173.1 3 - 4 398 56
GenBank mRNA BC134546.1 - 3 - 4 398 56

View Full Product Details