Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSR1
Entrez Gene ID
281311
Gene Name
macrophage scavenger receptor 1
Gene Aliases
-
Chromosome Location
Chr.27: 20909832 - 20990599 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113240.1 NP_001106711.1 2 - 3 142 106
NM_174113.2 NP_776538.1 2 - 3 142 106
GenBank mRNA BC142250.1 - 2 - 3 214 106
DY085614.1 - 2 - 3 209 106
X51689.1 - 1 - 1 117 106
X54183.1 - - 131 106

View Full Product Details