Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPX3
Entrez Gene ID
281210
Gene Name
glutathione peroxidase 3
Gene Aliases
-
Chromosome Location
Chr.7: 62284773 - 62292970 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174077.5 NP_776502.1 4 - 5 545 89
NR_138142.2 3 - 4 391 89
GenBank mRNA BC149266.1 - 4 - 5 524 89
DT832453.1 - 1 - 2 69 89
DT856720.1 - 4 - 5 522 89
DT861549.1 - 4 - 5 515 89
DV837910.1 - 4 - 5 523 89
DV838871.1 - 3 - 4 392 89
DV850873.1 - 3 - 4 370 89
EV673311.1 - 3 - 4 373 89
EV679018.1 - 4 - 5 525 89
EV688099.1 - 1 - 2 64 89
L10325.1 - 4 - 5 470 89

View Full Product Details