Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FST
Entrez Gene ID
327681
Gene Name
follistatin
Gene Aliases
-
Chromosome Location
Chr.20: 25581178 - 25586895 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175801.3 NP_786995.2 5 - 6 1129 68
GenBank mRNA BC133637.1 - 5 - 6 1130 68
L21716.1 - 5 - 6 951 68

View Full Product Details