Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF3M
Entrez Gene ID
515543
Gene Name
eukaryotic translation initiation factor 3 subunit M
Gene Aliases
HFL-B5
Chromosome Location
Chr.15: 63298831 - 63316295 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035312.2 NP_001030389.2 2 - 3 268 66
GenBank mRNA BC102130.1 - 2 - 3 237 66
DV802255.1 - 2 - 3 269 66

View Full Product Details