Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF3M
Entrez Gene ID
515543
Gene Name
eukaryotic translation initiation factor 3 subunit M
Gene Aliases
HFL-B5
Chromosome Location
Chr.15: 63298831 - 63316295 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035312.2 NP_001030389.2 5 - 6 623 85
GenBank mRNA BC102130.1 - 5 - 6 592 85
DV802255.1 - 5 - 6 624 85

View Full Product Details