Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HM13
Entrez Gene ID
512534
Gene Name
histocompatibility minor 13
Gene Aliases
-
Chromosome Location
Chr.13: 61107897 - 61146837 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046124.2 NP_001039589.2 10 - 11 1066 70
GenBank mRNA BC112454.1 - 10 - 11 1048 70

View Full Product Details