Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAQR6
Entrez Gene ID
516498
Gene Name
progestin and adipoQ receptor family member 6
Gene Aliases
-
Chromosome Location
Chr.3: 14545370 - 14551100 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046225.2 NP_001039690.2 1 - 2 125 127
GenBank mRNA EE359306.1 - 1 - 2 125 127

View Full Product Details