Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLMP
Entrez Gene ID
516565
Gene Name
CXADR like membrane protein
Gene Aliases
ASAM
Chromosome Location
Chr.15: 33689071 - 33794265 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098043.1 NP_001091512.1 4 - 5 870 104
GenBank mRNA BC140573.1 - 4 - 5 870 104

View Full Product Details