Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OR12D2E
Entrez Gene ID
510775
Gene Name
olfactory receptor family 12 subfamily D member 2E
Gene Aliases
OR12D2
Chromosome Location
Chr.23: 29305932 - 29311065 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098382.1 NP_001091852.1 1 - 2 304 120
GenBank mRNA BC134564.1 - 1 - 2 304 120

View Full Product Details