Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSTN
Entrez Gene ID
511114
Gene Name
osteocrin
Gene Aliases
-
Chromosome Location
Chr.1: 76065892 - 76102113 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098935.1 NP_001092405.1 1 - 2 54 87
GenBank mRNA BC142475.1 - 1 - 2 54 87

View Full Product Details