Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSTN
Entrez Gene ID
511114
Gene Name
osteocrin
Gene Aliases
-
Chromosome Location
Chr.1: 76065892 - 76102113 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098935.1 NP_001092405.1 3 - 4 371 76
GenBank mRNA BC142475.1 - 3 - 4 371 76
EF646361.1 - 2 - 3 326 76

View Full Product Details