Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC80
Entrez Gene ID
515235
Gene Name
coiled-coil domain containing 80
Gene Aliases
-
Chromosome Location
Chr.1: 57376258 - 57478031 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098982.2 NP_001092452.1 4 - 5 2906 101
GenBank mRNA BC142415.1 - 4 - 5 2907 101
BC151288.1 - 4 - 5 2654 101

View Full Product Details