Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PINK1
Entrez Gene ID
510683
Gene Name
PTEN induced kinase 1
Gene Aliases
-
Chromosome Location
Chr.2: 131986296 - 132006686 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099701.2 NP_001093171.1 2 - 3 714 82
GenBank mRNA BC142192.1 - 2 - 3 715 82

View Full Product Details