Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PI4K2A
Entrez Gene ID
507860
Gene Name
phosphatidylinositol 4-kinase type 2 alpha
Gene Aliases
-
Chromosome Location
Chr.26: 18819244 - 18845949 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100316.1 NP_001093786.1 1 - 2 449 78
GenBank mRNA BC148028.1 - 1 - 2 449 78

View Full Product Details