Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL34
Entrez Gene ID
508292
Gene Name
interleukin 34
Gene Aliases
C18H16ORF77, IL-34
Chromosome Location
Chr.18: 1510307 - 1570900 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100324.2 NP_001093794.1 3 - 4 697 77
GenBank mRNA BC146187.1 - 3 - 4 698 77

View Full Product Details