Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCRG1
Entrez Gene ID
515382
Gene Name
stimulator of chondrogenesis 1
Gene Aliases
-
Chromosome Location
Chr.8: 5783450 - 5890857 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100370.1 NP_001093840.1 1 - 2 82 106
GenBank mRNA BC149251.1 - 1 - 2 82 106
BC149633.1 - 1 - 2 85 106

View Full Product Details