Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC35G6
Entrez Gene ID
516150
Gene Name
solute carrier family 35 member G6
Gene Aliases
AMAC1L2, AMAC1L3
Chromosome Location
Chr.19: 27147404 - 27150342 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101095.1 NP_001094565.1 1 - 1 1551 63
GenBank mRNA EU025856.1 - 1 - 1 1551 63

View Full Product Details