Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDT9
Entrez Gene ID
517589
Gene Name
nudix hydrolase 9
Gene Aliases
-
Chromosome Location
Chr.6: 102320425 - 102357772 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101096.2 NP_001094566.1 1 - 2 353 124
GenBank mRNA BC151586.1 - 1 - 2 354 124

View Full Product Details