Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC13
Entrez Gene ID
511108
Gene Name
trafficking protein particle complex 13
Gene Aliases
C20H5orf44
Chromosome Location
Chr.20: 13902562 - 13938722 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101940.1 NP_001095410.1 4 - 5 565 152
GenBank mRNA BC151376.1 - 4 - 5 565 152

View Full Product Details