Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEPACAM2
Entrez Gene ID
513430
Gene Name
HEPACAM family member 2
Gene Aliases
-
Chromosome Location
Chr.4: 10540867 - 10600679 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101974.1 NP_001095444.1 9 - 10 1449 85
GenBank mRNA BC149765.1 - 9 - 10 1449 85

View Full Product Details