Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUBCNL
Entrez Gene ID
517231
Gene Name
rubicon like autophagy enhancer
Gene Aliases
C12H13orf18, KIAA0226L
Chromosome Location
Chr.12: 16369971 - 16410652 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102041.2 NP_001095511.1 3 - 4 762 118
GenBank mRNA BC151662.1 - 3 - 4 763 118

View Full Product Details