Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEH1L
Entrez Gene ID
506509
Gene Name
SEH1 like nucleoporin
Gene Aliases
-
Chromosome Location
Chr.24: 43196674 - 43216530 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103089.2 NP_001096559.1 3 - 4 414 108
GenBank mRNA BC151606.1 - 3 - 4 415 108

View Full Product Details