Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD8
Entrez Gene ID
520456
Gene Name
BTB (POZ) domain containing 8
Gene Aliases
-
Chromosome Location
Chr.3: 51222383 - 51316987 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105200.1 NP_001098670.1 2 - 3 347 130

View Full Product Details