Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERMARD
Entrez Gene ID
512804
Gene Name
ER membrane associated RNA degradation
Gene Aliases
C9H6orf70
Chromosome Location
Chr.9: 103548296 - 103577184 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105255.1 NP_001098725.1 13 - 14 1395 75

View Full Product Details