Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTSW
Entrez Gene ID
510967
Gene Name
cathepsin W
Gene Aliases
-
Chromosome Location
Chr.29: 43995837 - 43999310 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110070.1 NP_001103540.1 5 - 6 537 67
GenBank mRNA BC113312.1 - 5 - 6 537 67

View Full Product Details