Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCP11
Entrez Gene ID
523297
Gene Name
t-complex 11
Gene Aliases
-
Chromosome Location
Chr.23: 9078652 - 9239648 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110079.1 NP_001103549.1 5 - 6 612 87
GenBank mRNA BC111357.1 - 5 - 6 612 87

View Full Product Details