Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTS
Entrez Gene ID
506860
Gene Name
6-pyruvoyltetrahydropterin synthase
Gene Aliases
-
Chromosome Location
Chr.15: 22587044 - 22624477 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113537.1 NP_001107009.1 5 - 6 352 96
GenBank mRNA CB455463.1 - 5 - 6 357 96

View Full Product Details