Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRGPRF
Entrez Gene ID
615886
Gene Name
MAS related GPR family member F
Gene Aliases
-
Chromosome Location
Chr.29: 46354537 - 46364118 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035470.2 NP_001030547.2 1 - 2 179 69
GenBank mRNA BC118296.1 - 1 - 2 179 69
DT811968.1 - 1 - 2 134 69
DT814494.1 - 1 - 2 182 69

View Full Product Details